2,5-Dimethyl-3-hexyne-2,5-diol (±) + meso 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
2,5-Dimethylhex-3-yne-2,5-diol
B20889-22
B20889-36
CAAAAB20889-22EA
70.63
CAD
CAAAAB20889-22
CAAAAB20889-36
2,5-Dimethyl-3-hexyne-2,5-diol (±) + meso 98%
2,5-Dimethyl-3-hexyne-2,5-diol
Purity: 98%
CAS No.: 142-30-3
Molecular Formula: C8H14O2
Formula Weight: 142.20
Flash Point: 102°(215°F)
Melting Point: 90-94°
Boiling Point: 205°
Beilstein Reference: 969835
MDL No.: MFCD00004468
Formula:
C₈H₁₄O₂ MW: 142.2 g/mol Boiling Pt: 205 °C Melting Pt: 90…94 °C Density: 0.949 Flash Pt: 102 °C(215 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004468 CAS Number: 142-30-3 EINECS: 205-533-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...