4'-Cyclohexylacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Cyclohexylacetophenone

B20838-06 B20838-14 B20838-22
CAAAAB20838-06EA 42.03 CAD
CAAAAB20838-06 CAAAAB20838-14 CAAAAB20838-22
4'-Cyclohexylacetophenone 99%
4'-Cyclohexylacetophenone

Purity: 99% CAS No.: 18594-05-3 Molecular Formula: C14H18O Formula Weight: 202.30 Melting Point: 66-70° Boiling Point: 179-181°/14mm MDL No.: MFCD00001453

Formula: C₁₄H₁₈O
Boiling Pt: 179…181 °C/14 mm
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00001453
CAS Number: 18594-05-3
EINECS: 242-432-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR