Sodium trifluoromethanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20776-06
B20776-14
B20776-22
CAAAAB20776-06EA
91.96
CAD
CAAAAB20776-06
CAAAAB20776-14
CAAAAB20776-22
Sodium trifluoromethanesulfonate 98%
Sodium trifluoromethanesulphonate
Purity: 98%
CAS No.: 2926-30-9
Molecular Formula: CF3SO3Na
Formula Weight: 172.06
Melting Point: 251-255°
Beilstein Reference: 3728797
Formula:
CF₃NaO₃S Melting Pt: 251…255 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061607 CAS Number: 2926-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...