Diethyl fluoromalonate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20766-03
B20766-06
CAAAB20766-03EA
130.82
CAD
CAAAB20766-03
CAAAB20766-06
Diethyl fluoromalonate 97%
Diethyl fluoromalonate
Purity: 97%
CAS No.: 685-88-1
Molecular Formula: FCH(CO2CH2CH3)2
Formula Weight: 178.16
Flash Point: 62°(143°F)
Refractive Index: 1.4070
Boiling Point: 121-122°/30mm
Beilstein Reference: 1775686
UN No.: 1760
MDL No.: MFCD00009139
Formula:
C₇H₁₁FO₄ Boiling Pt: 121…122 °C/30 mm Density: 1.129 Flash Pt: 62 °C (143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009139 CAS Number: 685-88-1 EINECS: 211-684-7 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...