Ethylmalonic acid 98%
Supplier: Thermo Scientific Chemicals
B20715-06
B20715-14
B20715-22
CAAAB20715-06EA
69.84
CAD
CAAAB20715-06
CAAAB20715-14
CAAAB20715-22
Ethylmalonic acid 98%
Ethylmalonic acid
Purity: 98%
CAS No.: 601-75-2
Molecular Formula: CH3CH2CH(CO2H)2
Formula Weight: 132.12
Melting Point: 112-114°
Beilstein Reference: 774334
Formula:
C₅H₈O₄ Melting Pt: 112…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002668 CAS Number: 601-75-2 EINECS: 210-007-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...