1-Pentanesulfonic acid sodium salt, (max. 1% H₂O) 99% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

B20699-06 B20699-14 B20699-22
CAAAAB20699-06EA 85.95 CAD
CAAAAB20699-06 CAAAAB20699-14 CAAAAB20699-22
1-Pentanesulfonic acid sodium salt, (max. 1% H₂O) 99% (dry weight)
1-Pentanesulphonic acid sodium salt

Purity: 99% CAS No.: 22767-49-3 Molecular Formula: CH3(CH2)4SO3Na Formula Weight: 174.19 Melting Point: 300° Beilstein Reference: 3725147

Formula: C₅H₁₁O₃SNa
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 22767-49-3
EINECS: 245-208-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb1c23c8a6fe3c156a3aacb62721e1d1a34efb0&vid=1936dcdb47ba374ce21fd758660f7ca6ba95d254&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR