3,5-Dimethylanisole 99%
Supplier: Thermo Scientific Chemicals
Warning
|
B20651-14
B20651-22
CAAAAB20651-14EA
99.22
CAD
CAAAAB20651-14
CAAAAB20651-22
3,5-Dimethylanisole 99%
3,5-Dimethylanisole
Purity: 99%
CAS No.: 874-63-5
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 65°(149°F)
Refractive Index: 1.5130
Boiling Point: 192-193°
Beilstein Reference: 2040905
MDL No.: MFCD00008398
Formula:
C₉H₁₂O Boiling Pt: 192…193 °C Melting Pt: –0.2 °C Density: 0.960 Flash Pt: 65 °C(149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008398 CAS Number: 874-63-5 EINECS: 212-865-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...