Sodium hexafluoroantimonate 98%

Supplier: Thermo Scientific Chemicals
Warning

B20604-14 B20604-22 B20604-36
CAAAAB20604-14EA 83.42 CAD
CAAAAB20604-14 CAAAAB20604-22 CAAAAB20604-36
Sodium hexafluoroantimonate 98%
Sodium hexafluoroantimonate

Purity: 98% CAS No.: 16925-25-0 Molecular Formula: NaSbF6 Formula Weight: 258.73 Odor: Odorless UN No.: 1549 MDL No.: MFCD00003482

Formula: F₆NaSb
Density: 3.38
Storage Temperature: Ambient
MDL Number: MFCD00003482
CAS Number: 16925-25-0
EINECS: 240-989-8
UN: 1549
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR