2,3-Dihydrofuran 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20575-14
B20575-22
B20575-36
CAAAB20575-14EA
57.35
CAD
CAAAB20575-14
CAAAB20575-22
CAAAB20575-36
2,3-Dihydrofuran 98+%
2,3-Dihydrofuran
Purity: 98+%
CAS No.: 1191-99-7
Molecular Formula: C4H6O
Formula Weight: 70.09
Storage Temperature: 2°C to 8°C
Flash Point: -24°(-11°F)
Refractive Index: 1.4245
Odor: Pungent
Boiling Point: 54-55°
Beilstein Reference: 103168
UN No.: 3271
MDL No.: MFCD00003205
Formula:
C₄H₆O Boiling Pt: 54…55 °C Density: 0.927 Flash Pt: -24 °C(-11 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003205 CAS Number: 1191-99-7 EINECS: 214-747-7 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...