1-Acetyl-5-bromoindoline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20509-14
B20509-03
B20509-06
CAAAAB20509-14EA
910.08
CAD
CAAAAB20509-14
CAAAB20509-03
CAAAB20509-06
1-Acetyl-5-bromoindoline 98%
1-Acetyl-5-bromoindoline
Purity: 98%
CAS No.: 22190-38-1
Molecular Formula: C10H10BrNO
Formula Weight: 240.10
Melting Point: 118-121°
Beilstein Reference: 165052
Formula:
C₁₀H₁₀BrNO Melting Pt: 118…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00056017 CAS Number: 22190-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...