1-Hexene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20271-AE
B20271-AP
CAAAAB20271-AEEA
47.4
CAD
CAAAAB20271-AE
CAAAAB20271-AP
1-Hexene 98%
1-Hexene
Purity: 98%
CAS No.: 592-41-6
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -25°(-13°F)
Refractive Index: 1.3880
Odor: Mild
Melting Point: -140°
Boiling Point: 62-64°
Beilstein Reference: 1209240
UN No.: 2370
MDL No.: MFCD00009505
Formula:
C₆H₁₂ Boiling Pt: 62…64 °C Melting Pt: –140 °C Density: 0.673 Flash Pt: –25 °C (–13 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009505 CAS Number: 592-41-6 EINECS: 209-753-1 UN: 2370 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...