Trimethyl borate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20215-AK
B20215-AU
CAAAAB20215-AKEA
53.09
CAD
CAAAAB20215-AK
CAAAAB20215-AU
Trimethyl borate 99%
Trimethyl borate
Purity: 99%
CAS No.: 121-43-7
Molecular Formula: C3H9BO3
Formula Weight: 103.92
Flash Point: -8°(17°F)
Refractive Index: 1.3610
Odor: Odorless
Melting Point: -34°
Boiling Point: 67-69°
Beilstein Reference: 1697939
Merck Reference: 14,9712
UN No.: 2416
MDL No.: MFCD00008346
Formula:
C₃H₉BO₃ Boiling Pt: 67…69 °C Melting Pt: –34 °C Density: 0.915 Flash Pt: –8 °C (17 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008346 CAS Number: 121-43-7 EINECS: 204-468-9 UN: 2416 ADR: 3,II Merck Index: 14,9712 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...