Nitronium tetrafluoroborate 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20167-06
B20167-14
B20167-22
CAAAAB20167-06EA
102.7
CAD
CAAAAB20167-06
CAAAAB20167-14
CAAAAB20167-22
Nitronium tetrafluoroborate 96%
Nitronium tetrafluoroborate
Purity: 96%
CAS No.: 13826-86-3
Molecular Formula: BF4NO2
Formula Weight: 132.81
Storage Temperature: 2°C to 8°C
Melting Point: ca 240° dec.
UN No.: 3260
MDL No.: MFCD00011432
Formula:
BF₄NO₂ Melting Pt: ∼ 240 °C (dec.) Density: 1.26 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00011432 CAS Number: 13826-86-3 EINECS: 237-533-5 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...