2,4-Diamino-6-phenyl-1,3,5-triazine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20157-22
B20157-0E
B20157-36
CAAAAB20157-22EA
35.23
CAD
CAAAAB20157-22
CAAAB20157-0E
CAAAB20157-36
2,4-Diamino-6-phenyl-1,3,5-triazine 99%
2,4-Diamino-6-phenyl-1,3,5-triazine
Purity: 99%
CAS No.: 91-76-9
Molecular Formula: C9H9N5
Formula Weight: 187.21
Melting Point: 225-228°
Beilstein Reference: 153223
Merck Reference: 14,1089
MDL No.: MFCD00023187
Formula:
C₉H₉N₅ Melting Pt: 225…228 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023187 CAS Number: 91-76-9 EINECS: 202-095-6 Merck Index: 14,1089 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...