1-Bromo-4-(difluoromethoxy)benzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Difluoromethoxy)bromobenzene
B20106-06
B20106-14
CAAAAB20106-03EA
170.86
CAD
CAAAAB20106-03
CAAAAB20106-06
CAAAAB20106-14
1-Bromo-4-(difluoromethoxy)benzene 97%
1-Bromo-4-(difluoromethoxy)benzene
Purity: 97%
CAS No.: 5905-69-1
Molecular Formula: C7H5BrF2O
Formula Weight: 223.02
Refractive Index: 1.5010
Boiling Point: 96-98°/15mm
UN No.: 1993
MDL No.: MFCD00221465
Formula:
C₇H₅BrF₂O Boiling Pt: 96…98 °C/15 mm Density: 1.631 Storage Temperature: Ambient |
MDL Number:
MFCD00221465 CAS Number: 5905-69-1 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...