Tetradecylbenzene 97%

Supplier: Thermo Scientific Chemicals

A19988-06 A19988-14
CAAAAA19988-06EA 63.67 CAD
CAAAAA19988-06 CAAAAA19988-14
Tetradecylbenzene 97%
Tetradecylbenzene

Purity: 97% CAS No.: 1459-10-5 Molecular Formula: C6H5(CH2)13CH3 Formula Weight: 274.49 Refractive Index: 1.4856 Melting Point: 9° Boiling Point: 196°/6mm MDL No.: MFCD00015080

Formula: C₂₀H₃₄
MW: 274.49 g/mol
Boiling Pt: 196 °C (6 mmHg)
Melting Pt: 9 °C
Density: 0.8565
Storage Temperature: Ambient
MDL Number: MFCD00015080
CAS Number: 1459-10-5
EINECS: 215-950-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR