Sodium sulfate, anhydrous 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt

A19890-0B A19890-0I
CAAAAA19890-0BEA 89.27 CAD
CAAAAA19890-0B CAAAAA19890-0I
Sodium sulfate, anhydrous 99%
Sodium sulphate

Purity: 99% CAS No.: 7757-82-6 Molecular Formula: Na2O4S Formula Weight: 142.04 Refractive Index: 1.484 Odor: Odorless Merck Reference: 14,8680 MDL No.: MFCD00003504

Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429
Melting Pt: 884 °C
Density: 2.680
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 14,8680
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR