Sodium diphenylamine-4-sulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diphenylamine-4-sulfonic acid, sodium salt, Sodium Diphenylaminesulphonate, Sodium N-phenylsulphanilate, Sodium Diphenylaminesulfonate, Sodium (4-anilinobenzenesulfonate), Sodium (4-anilinobenzenesulphonate), Sodium N-phenylsulfanilate, Diphenylamine-4-sulphonic acid, sodium salt
A19768-06
A19768-14
CAAAAA19768-06EA
121.34
CAD
CAAAAA19768-06
CAAAAA19768-14
Sodium diphenylamine-4-sulfonate 98%
Sodium diphenylamine-4-sulphonate
Purity: 98%
CAS No.: 6152-67-6
Molecular Formula: C12H10NNaO3S
Formula Weight: 271.27
MDL No.: MFCD00036468
Formula:
C₁₂H₁₀NNaO₃S MW: 271.27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00036468 CAS Number: 6152-67-6 EINECS: 228-165-6 Merck Index: 13,07397 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...