2,2,4-Trimethylpentane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
A19756-0F
A19756-AP
CAAAAA19756-0FEA
156.74
CAD
CAAAAA19756-0F
CAAAAA19756-AP
2,2,4-Trimethylpentane 99%
2,2,4-Trimethylpentane
Purity: 99%
CAS No.: 540-84-1
Molecular Formula: C8H18
Formula Weight: 114.23
Flash Point: -7°(19°F)
Refractive Index: 1.3910
Odor: Nearly odorless
Melting Point: -107°
Boiling Point: 98-99°
Beilstein Reference: 1696876
Merck Reference: 14,5193
UN No.: 1262
MDL No.: MFCD00008943
Formula:
(CH₃)₂CHCH₂C(CH₃)₃ Boiling Pt: 98…99 °C Melting Pt: –107 °C Density: 0.692 Flash Pt: –7 °C (19 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008943 CAS Number: 540-84-1 EINECS: 208-759-1 UN: 1262 ADR: 3,II Merck Index: 14,5193 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...