6-Chlorooxindole 98%

Supplier: Thermo Scientific Chemicals
Warning

A19674-06 A19674-14 A19674-22
CAAAA19674-06EA 36.97 CAD
CAAAA19674-06 CAAAA19674-14 CAAAA19674-22
6-Chlorooxindole 98%
6-Chlorooxindole

Purity: 98% CAS No.: 56341-37-8 Molecular Formula: C8H6ClNO Formula Weight: 167.60 Melting Point: 195-199° MDL No.: MFCD00209962

Formula: C₈H₆ClNO
Melting Pt: 195…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00209962
CAS Number: 56341-37-8
EINECS: 421-330-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR