Phenanthrene tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19646-0E
A19646-22
A19646-36
CAAAAA19646-0EEA
1628.98
CAD
CAAAAA19646-0E
CAAAAA19646-22
CAAAAA19646-36
Phenanthrene tech. 90%, Technical Grade
Phenanthrene
MDL: MFCD00001168
Beilstein: 1905428
Soluble in alcohol, benzene, toluene, and glacial acetic acid
Purity: 90%
CAS No.: 85-01-8
Molecular Formula: C14H10
Formula Weight: 178.23
Flash Point: 171°(339°F)
Melting Point: 97-101°
Boiling Point: 340°
Solubility: Soluble in alcohol, benzene, toluene, and glacial acetic acid
Beilstein Reference: 1905428
Merck Reference: 14,7212
UN No.: 3077
MDL No.: MFCD00001168
Formula:
C₁₄H₁₀ Boiling Pt: 340 °C Melting Pt: 97…101 °C Density: 0.980 Flash Pt: 171 °C (339 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001168 CAS Number: 85-01-8 EINECS: 201-581-5 UN: 3077 ADR: 9,III Merck Index: 14,7212 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...