Bromoacetyl bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

A19580-14 A19580-22 A19580-36
CAAAAA19580-14EA 48.03 CAD
CAAAAA19580-14 CAAAAA19580-22 CAAAAA19580-36
Bromoacetyl bromide 98%
Bromoacetyl bromide

Purity: 98% CAS No.: 598-21-0 Molecular Formula: BrCH2COBr Formula Weight: 201.86 Flash Point: 105°(221°F) Refractive Index: 1.5460 Odor: Acrid Boiling Point: 147-150° Beilstein Reference: 605440 UN No.: 2513 MDL No.: MFCD00000115

Formula: BrCH₂COOBr
Boiling Pt: 147…150 °C
Density: 2.324
Flash Pt: >105 °C (221 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000115
CAS Number: 598-21-0
EINECS: 209-923-5
UN: 2513
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR