9-Decen-1-ol 90+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19490-14
A19490-22
CAAAAA19490-14EA
104.91
CAD
CAAAAA19490-14
CAAAAA19490-22
9-Decen-1-ol 90+%
9-Decen-1-ol
Purity: 90+%
CAS No.: 13019-22-2
Molecular Formula: C10H20O
Formula Weight: 156.27
Flash Point: 104°(219°F)
Refractive Index: 1.4480
Odor: Pleasant
Melting Point: -13°
Boiling Point: 235°
Beilstein Reference: 1750928
MDL No.: MFCD00002992
Formula:
C₁₀H₂₀O Boiling Pt: 235 °C Melting Pt: –13 °C Density: 0,847 Flash Pt: 104 °C(219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002992 CAS Number: 13019-22-2 EINECS: 235-878-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...