Butyl acetate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19412-0F
A19412-AP
CAAAAA19412-0FEA
203.82
CAD
CAAAAA19412-0F
CAAAAA19412-AP
Butyl acetate 99+%
Butyl acetate
Purity: 99+%
CAS No.: 123-86-4
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 25°(77°F)
Refractive Index: 1.3945
Odor: Fruit-like
pH: 5
Melting Point: -78°
Boiling Point: 124-126°
Beilstein Reference: 1741921
Merck Reference: 14,1535
UN No.: 1123
MDL No.: MFCD00009445
Formula:
CH₃COO(CH₂)₃CH₃ Boiling Pt: 124…126 °C Melting Pt: –78 °C Density: 0.881 Flash Pt: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009445 CAS Number: 123-86-4 EINECS: 204-658-1 UN: 1123 ADR: 3,III Merck Index: 14,1535 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...