1-Chloro-3-fluoro-4-iodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloro-1-fluoro-2-iodobenzene, 4-Chloro-2-fluoro-1-iodobenzene, 4-Chloro-2-fluoroiodobenzene

A19376-06 A19376-14
CAAAAA19376-06EA 89.43 CAD
CAAAAA19376-06 CAAAAA19376-14
1-Chloro-3-fluoro-4-iodobenzene 98%
1-Chloro-3-fluoro-4-iodobenzene

Purity: 98% CAS No.: 6797-79-1 Molecular Formula: C6H3ClFI Formula Weight: 256.45 Flash Point: 110°(230°F) Refractive Index: 1.6060 Boiling Point: 217° Beilstein Reference: 6288419 MDL No.: MFCD00079729

Formula: C₆H₃ClFI
MW: 256.45 g/mol
Boiling Pt: 217 °C
Density: 1.980
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00079729
CAS Number: 6797-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR