Ethyl phenylacetate 99%

Supplier: Thermo Scientific Chemicals
Warning

A19356-0E A19356-22 A19356-36
CAAAA19356-0EEA 360.24 CAD
CAAAA19356-0E CAAAA19356-22 CAAAA19356-36
Ethyl phenylacetate 99%
Ethyl phenylacetate
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 223…225 °C
Melting Pt: –29 °C
Density: 1.030
Flash Pt: 77 °C(170 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009178
CAS Number: 101-97-3
EINECS: 202-993-8
Merck Index: 14,3840

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR