3,4-Dimethylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A19260-14 A19260-22 A19260-36
CAAAAA19260-14EA 73.94 CAD
CAAAAA19260-14 CAAAAA19260-22 CAAAAA19260-36
3,4-Dimethylbenzoic acid 98%
3,4-Dimethylbenzoic acid

Purity: 98% CAS No.: 619-04-5 Molecular Formula: C9H10O2 Formula Weight: 150.18 Odor: Odorless Melting Point: 164-167° Beilstein Reference: 907267 MDL No.: MFCD00002524

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00002524
CAS Number: 619-04-5
EINECS: 210-576-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR