5-Chlorooxindole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Chloro-2-oxindole
A19210-06
A19210-14
CAAAA19210-06EA
316
CAD
CAAAA19210-06
CAAAA19210-14
5-Chlorooxindole 98%
5-Chlorooxindole
Purity: 98%
CAS No.: 17630-75-0
Molecular Formula: C8H6ClNO
Formula Weight: 167.60
Melting Point: 196-200°
MDL No.: MFCD00191927
Formula:
C₈H₆ClNO MW: 167.59 g/mol Melting Pt: 196…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191927 CAS Number: 17630-75-0 EINECS: 412-200-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...