Potassium hydroxide ≥85%, pellets
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18854-0E
A18854-36
CAAAAA18854-0EEA
161.16
CAD
CAAAAA18854-0E
CAAAAA18854-36
Potassium hydroxide ≥85%, pellets
Potassium hydroxide
Purity: 85%
CAS No.: 1310-58-3
Molecular Formula: KOH
Formula Weight: 56.11
Odor: Odorless
pH: 13,5
Melting Point: 360°
Boiling Point: 1320-1324°
Merck Reference: 14,7640
UN No.: 1813
MDL No.: MFCD00003553
Formula:
KOH Boiling Pt: 1320…1324 °C Melting Pt: 360 °C Density: 2.044 Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 1813 ADR: 8,II REACH: 01-2119487136-33 Merck Index: 14,7640 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...