Manganese (II) acetylacetonate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(2,4-pentanedionato-O,O')manganese (II), Manganese (II) bis(2,4-pentanedionato-O,O'), Manganese(II) bis(acetylacetonate), Manganese (II) 2,4-pentanedionate

A18762-14 A18762-22 A18762-36
CAAAAA18762-14EA 56.25 CAD
CAAAAA18762-14 CAAAAA18762-22 CAAAAA18762-36
Manganese (II) acetylacetonate
Manganese (II) acetylacetonate

CAS No.: 14024-58-9 Molecular Formula: Mn(C5H7O2)2 Formula Weight: 253.16 Melting Point: 260-262° dec. MDL No.: MFCD00000022

Formula: C₁₀H₁₄MnO₄
MW: 253.16 g/mol
Melting Pt: 260…262 °C (dec.)
Density: 1.60
Storage Temperature: Ambient
MDL Number: MFCD00000022
CAS Number: 14024-58-9
EINECS: 237-858-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR