4-Nitrophenyl chloroformate 97%

Supplier: Thermo Scientific Chemicals
Danger

A18742-06 A18742-14 A18742-22
CAAAAA18742-06EA 70.94 CAD
CAAAAA18742-06 CAAAAA18742-14 CAAAAA18742-22
4-Nitrophenyl chloroformate 97%
4-Nitrophenyl chloroformate

Purity: 97% CAS No.: 7693-46-1 Molecular Formula: C7H4ClNO4 Formula Weight: 201.57 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Odor: Acrid Melting Point: 75-82° Boiling Point: 159-162°/19mm Beilstein Reference: 518127 UN No.: 3261 MDL No.: MFCD00007321

Formula: C₇H₄ClNO₄
Boiling Pt: 159…162 °C/19 mm
Melting Pt: 75…82 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007321
CAS Number: 7693-46-1
EINECS: 231-706-9
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR