Sulfamide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sulfuric diamide, Sulphuric diamide

A18680-09 A18680-18 A18680-88
CAAAA18680-09EA 170.64 CAD
CAAAA18680-09 CAAAA18680-18 CAAAA18680-88
Sulfamide 99%
Sulfamide

MDL: MFCD00011606 Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol


Purity: 99% CAS No.: 7803-58-9 Molecular Formula: SO2(NH2)2 Formula Weight: 96.11 Melting Point: 91-92° Solubility: Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol MDL No.: MFCD00011606

Formula: H₄N₂O₂S
MW: 96.11
Melting Pt: 91…92 °C
Density: 1.61
Storage Temperature: Ambient
MDL Number: MFCD00011606
CAS Number: 7803-58-9
EINECS: 232-262-9
Merck Index: 14,8922

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR