Triphenyl phosphite 97%

Supplier: Thermo Scientific Chemicals
Warning

A18662-0E A18662-36
CAAAAA18662-0EEA 252.8 CAD
CAAAAA18662-0E CAAAAA18662-36
Triphenyl phosphite 97%
Triphenyl phosphite

Purity: 97% CAS No.: 101-02-0 Molecular Formula: C18H15O3P Formula Weight: 310.29 Flash Point: 146°(295°F) Odor: Mild Boiling Point: 360° UN No.: 3077 MDL No.: MFCD00003032

Formula: C₁₈H₁₅O₃P
MW: 310.29 g/mol
Boiling Pt: 360 °C
Melting Pt: 22…24 °C
Density: 1.184
Flash Pt: 146 °C (295 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003032
CAS Number: 101-02-0
EINECS: 202-908-4
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR