Sodium stannate trihydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium tin(IV) oxide trihydrate, Disodium tin trioxide trihydrate

A18611-0E A18611-22 A18611-36
CAAAAA18611-0EEA 546.68 CAD
CAAAAA18611-0E CAAAAA18611-22 CAAAAA18611-36
Sodium stannate trihydrate 98%
Sodium stannate trihydrate

Purity: 98% CAS No.: 12209-98-2 Molecular Formula: Na2SnO3.3H2O Formula Weight: 266.75 (212.67anhy) Odor: Odorless Merck Reference: 14,8677 MDL No.: MFCD00150189

Formula: Na₂O₃Sn·3H₂O
MW: 266.73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150189
CAS Number: 12209-98-2
EINECS: 235-030-5
Merck Index: 14,8677

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR