3-Fluoro-p-anisidine 98+%

Supplier: Thermo Scientific Chemicals
Warning

A18606-14 A18606-06
CAAAAA18606-14EA 824.76 CAD
CAAAAA18606-14 CAAAA18606-06
3-Fluoro-p-anisidine 98+%
3-Fluoro-p-anisidine

Purity: 98+% CAS No.: 366-99-4 Molecular Formula: C7H8FNO Formula Weight: 141.15 Flash Point: 110°(230°F) Melting Point: 80-84° Boiling Point: 135°/18mm Beilstein Reference: 2717083 UN No.: 2811 MDL No.: MFCD00075040

Formula: C₇H₈FNO
Boiling Pt: 135 °C/18 mm
Melting Pt: 80…84 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075040
CAS Number: 366-99-4
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR