Ammonium hydrogen carbonate 98%

Supplier: Thermo Scientific Chemicals
Warning

A18566-0E A18566-36
CAAAAA18566-0EEA 129.88 CAD
CAAAAA18566-0E CAAAAA18566-36
Ammonium hydrogen carbonate 98%
Ammonium hydrogen carbonate

Purity: 98% CAS No.: 1066-33-7 Molecular Formula: (NH4)HCO3 Formula Weight: 79.06 Odor: Like ammonia Melting Point: ca 60° dec. Beilstein Reference: 4329606 Merck Reference: 14,497 MDL No.: MFCD00012138

Formula: NH₄HCO₃
Melting Pt: ∼ 60 °C (dec.)
Density: 1.586
Storage Temperature: Ambient
MDL Number: MFCD00012138
CAS Number: 1066-33-7
EINECS: 213-911-5
Merck Index: 14,497

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR