Manganese(II) nitrate tetrahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Manganous nitrate tetrahydrate, Manganese dinitrate tetrahydrate

A18521-0B A18521-30
CAAAAA18521-0BEA 301.78 CAD
CAAAAA18521-0B CAAAAA18521-30
Manganese(II) nitrate tetrahydrate 98%
Manganese (II) nitrate tetrahydrate

Purity: 98% CAS No.: 20694-39-7 Molecular Formula: MnN2O6.4H2O Formula Weight: 251.01 (178.95anhy) Odor: Odorless Melting Point: ca 26° UN No.: 2724 MDL No.: MFCD00149789

Formula: Mn(NO₃)₂·4H₂O
Melting Pt: ∼ 26 °C
Density: 2.115 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149789
CAS Number: 20694-39-7
EINECS: 233-828-8
UN: 2724
ADR: 5.1,III
Merck Index: 14,5733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR