Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
A18494-0E
A18494-22
A18494-36
CAAAAA18494-0EEA
357.08
CAD
CAAAAA18494-0E
CAAAAA18494-22
CAAAAA18494-36
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99%
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Purity: 99%
CAS No.: 77-86-1
Molecular Formula: C4H11NO3
Formula Weight: 121.14
Melting Point: 168-172°
Boiling Point: 219-220°/10mm
Merck Reference: 14,9772
MDL No.: MFCD00004679
Formula:
NH₂C(CH₂OH)₃ MW: 121.14 g/mol Boiling Pt: 357 Melting Pt: 168…172 °C Density: 1.353 Storage Temperature: Ambient |
MDL Number:
MFCD00004679 CAS Number: 77-86-1 EINECS: 201-064-4 Merck Index: 14,9772 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...