2-Amino-4-phenylthiazole 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Phenylthiazol-2-amine, 4-Phenyl-1,3-thiazol-2-amine
A18488-03
A18488-06
CAAAA18488-03EA
60.36
CAD
CAAAA18488-03
CAAAA18488-06
2-Amino-4-phenylthiazole 98%
2-Amino-4-phenylthiazole
Purity: 98%
CAS No.: 2010-06-2
Molecular Formula: C9H8N2S
Formula Weight: 176.24
Odor: Odorless
Melting Point: 148-152°
Beilstein Reference: 128609
UN No.: 2811
MDL No.: MFCD00039680
Formula:
C₉H₈N₂S Melting Pt: 148…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039680 CAS Number: 2010-06-2 EINECS: 217-926-8 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...