Germanium(IV) methoxide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18437-06
A18437-14
CAAAAA18437-06EA
274.92
CAD
CAAAAA18437-06
CAAAAA18437-14
Germanium(IV) methoxide 98%
Germanium (IV) methoxide
Purity: 98%
CAS No.: 992-91-6
Molecular Formula: C4H12GeO4
Formula Weight: 196.73
Flash Point: 40°(105°F)
Refractive Index: 1.4015
Odor: Like alcohol
Melting Point: -18°
Boiling Point: 145°
UN No.: 2929
MDL No.: MFCD00014881
Formula:
C₄H₁₂GeO₄ Boiling Pt: 145 °C Melting Pt: –18 °C Density: 1.3254^2^0 Flash Pt: 40 °C (105 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014881 CAS Number: 992-91-6 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...