Nonafluoro-2-(trifluoromethyl)-2-pentene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Perfluoro(2-methyl-2-pentene), Perfluoro(2-methylpent-2-ene), 1,1,1,3,4,4,5,5,5-Nonafluoro-2-(trifluoromethyl)-2-pentene

A18394-06 A18394-14
CAAAAA18394-03EA 72.66 CAD
CAAAAA18394-03 CAAAAA18394-06 CAAAAA18394-14
Nonafluoro-2-(trifluoromethyl)-2-pentene 97%
Nonafluoro-2-(trifluoromethyl)-2-pentene

Purity: 97% CAS No.: 1584-03-8 Molecular Formula: (CF3)2C=CFCF2CF3 Formula Weight: 300.05 Flash Point: -12°(10°F) Refractive Index: ca. 1.3 Boiling Point: 48-54° UN No.: 1993 MDL No.: MFCD00015724

Formula: C₆F₁₂
MW: 300.05 g/mol
Boiling Pt: 48…54 °C
Density: 1.620
Flash Pt: –12 °C (10 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015724
CAS Number: 1584-03-8
EINECS: 216-436-1
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR