Cobalt(II) acetate tetrahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A18374-14 A18374-22 A18374-36
CAAAAA18374-14EA 45.03 CAD
CAAAAA18374-14 CAAAAA18374-22 CAAAAA18374-36
Cobalt(II) acetate tetrahydrate 98%
Cobalt (II) acetate tetrahydrate

Purity: 98% CAS No.: 6147-53-1 Molecular Formula: C4H6CoO4.4H2O Formula Weight: 249.08 (177.02anhy) Refractive Index: 1.542 Odor: Odorless Beilstein Reference: 3731397 Merck Reference: 14,2433 UN No.: 3077 MDL No.: MFCD00149569

Formula: (H₃CCOO)₂Co·4H₂O
Melting Pt: ∼ 140 °C (–H₂O of crystall.)
Density: 1.71
Storage Temperature: Ambient
MDL Number: MFCD00149569
CAS Number: 6147-53-1
EINECS: 200-755-8
UN: 3077
ADR: 9,III
Merck Index: 14,2433

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR