Diphenyl selenide 98%

Supplier: Thermo Scientific Chemicals
Danger

A18364-03 A18364-06 A18364-14
CAAAAA18364-03EA 52.93 CAD
CAAAAA18364-03 CAAAAA18364-06 CAAAAA18364-14
Diphenyl selenide 98%
Diphenyl selenide

Purity: 98% CAS No.: 1132-39-4 Molecular Formula: (C6H5)2Se Formula Weight: 233.17 Odor: Unpleasant Melting Point: 3° Boiling Point: 165-167°/12mm UN No.: 3440 MDL No.: MFCD00014067

Formula: C₁₂H₁₀Se
MW: 233.17
Boiling Pt: 165…167 °C (12 mmHg)
Melting Pt: 3 °C
Density: 1.351
Storage Temperature: Ambient
MDL Number: MFCD00014067
CAS Number: 1132-39-4
EINECS: 214-474-2
UN: 2810
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR