Manganese(II) bromide, anhydrous 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Manganous bromide, Manganese(ii)bromide, Manganese dibromide, Manganese(II) bromide hydrate

A18321-14 A18321-22
CAAAAA18321-14EA 140.46 CAD
CAAAAA18321-14 CAAAAA18321-22
Manganese(II) bromide, anhydrous 95%
Manganese (II) bromide

Purity: 95% CAS No.: 13446-03-2 Molecular Formula: MnBr2 Formula Weight: 214.76 Odor: Odorless Melting Point: 698° MDL No.: MFCD00011113

Formula: Br₂Mn
MW: 214.75 g/mol
Melting Pt: 698
Density: 4.38
Storage Temperature: Ambient
MDL Number: MFCD00011113
CAS Number: 13446-03-2
EINECS: 236-591-9
Merck Index: 14,5725

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR