Disodium hydrogenarsenate heptahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A18275-22 A18275-36
CAAAAA18275-22EA 134.93 CAD
CAAAAA18275-22 CAAAAA18275-36
Disodium hydrogenarsenate heptahydrate 98%
Disodium hydrogenarsenate heptahydrate

Purity: 98% CAS No.: 10048-95-0 Molecular Formula: Na2HAsO4.7H2O Formula Weight: 312.01 (185.94anhy) Odor: Odorless Melting Point: 130° dec. UN No.: 1685 MDL No.: MFCD00149179

Formula: HAsNa₂O₄·7H₂O
Melting Pt: 130 °C (dec.)
Density: 1.88
Storage Temperature: Ambient
MDL Number: MFCD00149179
CAS Number: 10048-95-0
EINECS: 231-902-4
UN: 1685
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR