Calcium-2-ethylhexanoate 98%

Supplier: Thermo Scientific Chemicals
Warning

A18266-06 A18266-14 A18266-22
CAAAAA18266-06EA 88.48 CAD
CAAAAA18266-06 CAAAAA18266-14 CAAAAA18266-22
Calcium-2-ethylhexanoate 98%
Calcium 2-ethylhexanoate

Purity: 98% CAS No.: 136-51-6 Molecular Formula: C16H30CaO4 Formula Weight: 326.50 Odor: Odorless

Formula: C₁₆H₃₀CaO₄
Storage Temperature: Ambient
MDL Number: MFCD00014001
CAS Number: 136-51-6
EINECS: 205-249-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR