Potassium bromate 99%

Supplier: Thermo Scientific Chemicals
Danger

A18258-0E A18258-22 A18258-36
CAAAAA18258-0EEA 576.7 CAD
CAAAAA18258-0E CAAAAA18258-22 CAAAAA18258-36
Potassium bromate 99%
Potassium bromate

Purity: 99% CAS No.: 7758-01-2 Molecular Formula: KBrO3 Formula Weight: 167.00 Odor: Odorless Melting Point: ca 370° dec. UN No.: 1484 MDL No.: MFCD00011359

Formula: KBrO₃
Melting Pt: ∼ 370 °C (dec.)
Density: 3.27
Storage Temperature: Ambient
MDL Number: MFCD00011359
CAS Number: 7758-01-2
EINECS: 231-829-8
UN: 1484
ADR: 5.1,II
Merck Index: 14,7617

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR