Indium(III) nitrate hydrate In ≥ 29%

Supplier: Thermo Scientific Chemicals
Danger

A18252-14 A18252-22
CAAAAA18252-14EA 205.4 CAD
CAAAAA18252-14 CAAAAA18252-22
Indium(III) nitrate hydrate In ≥ 29%
Indium (III) nitrate hydrate

CAS No.: 207398-97-8 Molecular Formula: In(NO3)3.xH2O Formula Weight: 300.83(anhy) Melting Point: 100° -H2O UN No.: 1477 MDL No.: MFCD00149742

Formula: InN₃O₉·H₂O
Melting Pt: 100 °C (–H₂O of crystall.)
Storage Temperature: Ambient
MDL Number: MFCD00149742
CAS Number: 207398-97-8
EINECS: 237-393-5
UN: 1477
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR