2,2,2-Trifluoroethyl acrylate 98% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2,2-Trifluoroethylacrylate, Acrylic acid 2,2,2-trifluoroethyl ester
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) 200 ppm
A18235-06
A18235-14
CAAAA18235-06EA
62.73
CAD
CAAAA18235-06
CAAAA18235-14
2,2,2-Trifluoroethyl acrylate 98% stabilised
2,2,2-Trifluoroethyl acrylate
Purity: 98%
CAS No.: 407-47-6
Molecular Formula: C5H5F3O2
Formula Weight: 154.09
Storage Temperature: 2°C to 8°C
Flash Point: 12°(53°F)
Refractive Index: 1.3500
Odor: Pungent
Boiling Point: 91-93°
Beilstein Reference: 1756611
UN No.: 3272
MDL No.: MFCD00000444
Formula:
C₅H₅F₃O₂ Boiling Pt: 91…93 °C Density: 1.216 Flash Pt: 12 °C(53 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000444 CAS Number: 407-47-6 EINECS: 206-987-6 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...