1-Hexanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18232-0F
A18232-AP
CAAAAA18232-0FEA
171.94
CAD
CAAAAA18232-0F
CAAAAA18232-AP
1-Hexanol 99%
1-Hexanol
Purity: 99%
CAS No.: 111-27-3
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 60°(140°F)
Physical Form: Clear liquid
Density: 0.814
Refractive Index: 1.4180
Appearance: Colorless
Melting Point: -52°
Boiling Point: 157°
Beilstein Reference: 969167
Merck Reference: 14,4697
UN No.: 2282
MDL No.: MFCD00002982
Formula:
H₃C(CH₂)₄CH₂OH Boiling Pt: 157 °C Melting Pt: –52 °C Density: 0.814 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002982 CAS Number: 111-27-3 EINECS: 203-852-3 UN: 2282 ADR: 3,III Merck Index: 14,4697 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...